Skip to Content
Merck
All Photos(1)

Key Documents

728608

Sigma-Aldrich

2′-Iodoacetanilide

≥97.0% (HPLC)

Synonym(s):

N-(2-Iodophenyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8INO
CAS Number:
Molecular Weight:
261.06
Beilstein:
2803067
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

form

powder

functional group

amide
iodo

SMILES string

CC(=O)Nc1ccccc1I

InChI

1S/C8H8INO/c1-6(11)10-8-5-3-2-4-7(8)9/h2-5H,1H3,(H,10,11)

InChI key

BCJOKHQYEDXBSF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2,3-disubstituted indoles via palladium-catalyzed annulation of internal alkynes.
Larock, R. C., et al.
The Journal of Organic Chemistry, 63(22), 7652-7662 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service