Skip to Content
Merck
All Photos(1)

Key Documents

728519

Sigma-Aldrich

3-(1-Piperazinyl)-1,2-benzisothiazole

≥98% (HPLC)

Synonym(s):

3-Piperazin-1-yl-benzo[d]isothiazole

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
₹14,786.95

₹14,786.95


Available to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
₹14,786.95

About This Item

Empirical Formula (Hill Notation):
C11H13N3S
CAS Number:
Molecular Weight:
219.31
Beilstein:
4253627
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

₹14,786.95


Available to ship on11 April 2025Details


Request a Bulk Order

Assay

≥98% (HPLC)

form

crystals

storage temp.

2-8°C

SMILES string

C1CN(CCN1)c2nsc3ccccc23

InChI

1S/C11H13N3S/c1-2-4-10-9(3-1)11(13-15-10)14-7-5-12-6-8-14/h1-4,12H,5-8H2

InChI key

KRDOFMHJLWKXIU-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service