Skip to Content
Merck
All Photos(1)

Key Documents

726346

Sigma-Aldrich

(R,S)-3-Benzyloxy-2-methylpropionaldehyde

≥95%

Synonym(s):

(R,S)-3-(Benzyloxy)-2-methylpropanal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O2
CAS Number:
Molecular Weight:
178.23
Beilstein:
2504211
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%
≥95.0% (GC)

form

liquid

storage temp.

−20°C

SMILES string

CC(COCc1ccccc1)C=O

InChI

1S/C11H14O2/c1-10(7-12)8-13-9-11-5-3-2-4-6-11/h2-7,10H,8-9H2,1H3

InChI key

XYUPRHVYEKOWCD-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service