Skip to Content
Merck
All Photos(2)

Key Documents

721239

Sigma-Aldrich

2,2′-Dinitro-4,4′-stilbenedicarboxylic acid

greener alternative

97%

Synonym(s):

H2DNSDC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10N2O8
CAS Number:
Molecular Weight:
358.26
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

248-252 °C

greener alternative category

SMILES string

OC(=O)c1ccc(\C=C\c2ccc(cc2[N+]([O-])=O)C(O)=O)c(c1)[N+]([O-])=O

InChI

1S/C16H10N2O8/c19-15(20)11-5-3-9(13(7-11)17(23)24)1-2-10-4-6-12(16(21)22)8-14(10)18(25)26/h1-8H,(H,19,20)(H,21,22)/b2-1+

InChI key

LSIRFWJQXQJPKC-OWOJBTEDSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

2,2′-Dinitro-4,4′-stilbenedicarboxylic acid can be used as an organic linker in the synthesis of metal-organic frameworks (MOFs) for potential application in the capture of CO2. The MOFs synthesized improve the adsorption of CO2 and characteristics like selectivity.[1]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Engineering Accessible Adsorption Sites in MOFs for CO2 Capture
Ingram C and Tandabany D
Advanced Sustainable Systems (2019)

Articles

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service