Skip to Content
Merck
All Photos(1)

Key Documents

704997

Sigma-Aldrich

Benzene-1,4-diboronic acid mono-MIDA ester

Synonym(s):

Benzene-1-boronic acid-4-boronic acid MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13B2NO6
CAS Number:
Molecular Weight:
276.85
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

mp

170-175 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2ccc(cc2)B(O)O

InChI

1S/C11H13B2NO6/c1-14-6-10(15)19-13(20-11(16)7-14)9-4-2-8(3-5-9)12(17)18/h2-5,17-18H,6-7H2,1H3

InChI key

RXUGTOKUNBZXQV-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service