Skip to Content
Merck
All Photos(1)

Documents

704911

Sigma-Aldrich

N-Benzyl-2-bromo-N-phenylbutanamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18BrNO
CAS Number:
Molecular Weight:
332.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

44-50 °C

SMILES string

CCC(Br)C(=O)N(Cc1ccccc1)c2ccccc2

InChI

1S/C17H18BrNO/c1-2-16(18)17(20)19(15-11-7-4-8-12-15)13-14-9-5-3-6-10-14/h3-12,16H,2,13H2,1H3

InChI key

DVAAHBQCAZCHOP-UHFFFAOYSA-N

Application

Used in Ni-pybox catalyzed asymmetric Negishi couplings.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Fischer et al.
Journal of the American Chemical Society, 127(13), 4594-4595 (2005-03-31)
A Ni/Pybox catalyst achieves the asymmetric cross-coupling of secondary alpha-bromo amides with organozinc reagents. The process tolerates a variety of functional groups and affords the desired product in good yield and in high enantiomeric excess.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service