Skip to Content
Merck
All Photos(1)

Key Documents

704040

Sigma-Aldrich

3-(Bromoacetyl)pyridine hydrobromide

95%

Synonym(s):

2-Bromo-1-(3-pyridyl)ethanone hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H6BrNO · HBr
CAS Number:
Molecular Weight:
280.94
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

solid

mp

192-197 °C

functional group

bromo
ketone

SMILES string

Br.BrCC(=O)c1cccnc1

InChI

1S/C7H6BrNO.BrH/c8-4-7(10)6-2-1-3-9-5-6;/h1-3,5H,4H2;1H

InChI key

WDTSYONULAZKIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Ulmer et al.
European journal of biochemistry, 136(1), 183-194 (1983-10-17)
Bromoacetylpyridine acts as an active-site-directed inhibitor on glucose dehydrogenase from Bacillus megaterium. The inactivation is irreversible with a Ki of 7.7 mM. The coenzyme NAD but not the substrate glucose protects the enzyme from the inactivation. It is proposed that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service