Skip to Content
Merck
All Photos(2)

Key Documents

701262

Sigma-Aldrich

2-Bromo-2-methylpropionamide

97%

Synonym(s):

2-Bromo-2,2-dimethylacetamide, 2-Bromo-2-methylpropanamide, 2-Bromoisobutyramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8BrNO
CAS Number:
Molecular Weight:
166.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

145-149 °C

functional group

amide
bromo

SMILES string

CC(C)(Br)C(N)=O

InChI

1S/C4H8BrNO/c1-4(2,5)3(6)7/h1-2H3,(H2,6,7)

InChI key

WTRIMJTZOOLIFZ-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francis Perton et al.
Journal of colloid and interface science, 542, 469-482 (2019-02-18)
The engineering of luminescent nanoplatforms for biomedical applications displaying ability for scaling-up, good colloidal stability in aqueous solutions, biocompatibility, and providing an easy detection in vivo by fluorescence methods while offering high potential of functionalities, is currently a challenge. The
Qian Chen et al.
ACS nano, 9(5), 5223-5233 (2015-05-08)
Paclitaxel (PTX) can bind to human serum albumin (HSA) via hydrophobic interaction, forming Abraxane, which is a U.S. Food and Drug Administration (FDA) approved effective antitumor nanomedicine drug. Herein, the effective antitumor drug PTX is used to induce the self-assembly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service