Skip to Content
Merck
All Photos(1)

Key Documents

696579

Sigma-Aldrich

2-Fluoro-3-pyridineboronic acid

≥95%

Synonym(s):

(2-Fluoropyridin-3-yl)boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5BFNO2
CAS Number:
Molecular Weight:
140.91
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

mp

274-283 °C

SMILES string

OB(O)c1cccnc1F

InChI

1S/C5H5BFNO2/c7-5-4(6(9)10)2-1-3-8-5/h1-3,9-10H

InChI key

YUHZIUAREWNXJT-UHFFFAOYSA-N

Application

Reactant involved in:
  • Heterocyclization eith α-oxocarboxylic acids
  • Suzuki-Miyaura coupling reactions

Additionally used as a precursor to biologically active molecules including:
  • Heteroaryl benzylureas with glycogen synthase kinase 3 inhibitory activity
  • Carboxyindoles with HCV NS5B polymerase inhibitory activity
  • Pyrazolyl- and thienyl-aminohydantoins with BACE1 inhibitory activity

Other Notes

May contain varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service