Skip to Content
Merck
All Photos(1)

Documents

696196

Sigma-Aldrich

1,3-Bis(2,6-diisopropylphenyl)-1,3-dihydro-2H-imidazol-2-ylidene

97%

Synonym(s):

Ipr, 1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36N2
CAS Number:
Molecular Weight:
388.59
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: ligand

mp

213-217 °C

storage temp.

−20°C

SMILES string

CC(C)c1cccc(C(C)C)c1N2[C]N(C=C2)c3c(cccc3C(C)C)C(C)C

InChI

1S/C27H36N2/c1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8/h9-16,18-21H,1-8H3

InChI key

VYCIHDBIKGRENI-UHFFFAOYSA-N

Application

Ligand for Pd complexes for amination reaction of aryl halides; used as C-C bond formation reaction, e.g. Kumada-Tamao-Corriu reaction, Suzuki coupling, and Stille coupling.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunxia Du et al.
Biochemical and biophysical research communications, 503(1), 177-180 (2018-06-11)
It is high incidence of tubulointerstitial lesion (TIL) in lupus nephritis (LN) and TIL can affect the prognosis of patients with LN. Signal transducer and activator of transcription (STAT) 3 was activated in LN and STAT3 inhibition could delay the
C Wen et al.
Transplantation proceedings, 47(8), 2523-2527 (2015-11-01)
For patients with refractory systemic lupus erythematosus, current medications are insufficient to control their condition, and new treatments are necessary. We aimed to evaluate the therapeutic effect of fetal and neonatal murine peripheral blood (FNPB) mononuclear cells and their impact
Sheng Gao et al.
Molecular immunology, 81, 127-134 (2016-12-13)
Over activation of conventional dendritic cells (cDCs) contributes to the development of systemic lupus erythematosus (SLE). Triggering receptor expressed on myeloid cells 1 (TREM-1) is emerging as a potent amplifier of the inflammatory responses. We sought to determine the expression
Zhang, C.; Huang, J. et al.
The Journal of Organic Chemistry, 64, 3804-3804 (1999)
H Stetsun et al.
Arthropod structure & development, 52, 100882-100882 (2019-09-01)
Digger wasps of the family Crabronidae (Insecta: Hymenoptera) are generally known to use their sting to paralyze or kill a prey. However, only a few species of digger wasps transport their prey to the nest impaled on the sting. How

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service