Recommended Products
General description
sold in collaboration with Solvias AG
Kit Components Also Available Separately
Product No.
Description
SDS
- 73463(SP,S′P)-1,1′-Bis[(R)-α-(dimethylamino)benzyl]-2,2′-bis(diphenylphosphino)ferrocene, ≥97% 100 mgSDS
- (SP,S′P)-1,1′-Bis(dicyclohexylphosphino)-2,2′-bis[(R)-α-(dimethylamino)benzyl]ferrocene, ≥97% 100 mg
- 73467(SP,S′P)-1,1′-Bis{bis[3,5-bis(trifluoromethyl)phenyl]phosphino}-2,2′-bis[(R)-α-(dimethylamino)benzyl]ferrocene, ≥97% 100 mgSDS
- 73469(SP,S′P)-1,1′-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2,2′-bis[(R)-α-(dimethylamino)benzyl]ferrocene, ≥97% 100 mgSDS
- 73471(SP,S′P)-1,1′-Bis[(R)-α-(dimethylamino)benzyl]-2,2′-bis[di(3,5-xylyl)phosphino]ferrocene, ≥97% 100 mgSDS
- (SP,S′P)-1,1′-Bis[bis(2-methylphenyl)phosphino]-2,2′-bis[(R)-α-(dimethylamino)benzyl]ferrocene, ≥97% 100 mg
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service