Skip to Content
Merck
All Photos(1)

Key Documents

690341

Sigma-Aldrich

2,3,4,6-Tetra-O-benzyl-D-gluconic acid-δ-lactone

Synonym(s):

(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-(benzyloxymethyl)tetrahydropyran-2-one, 2,3,4,6-Tetra-O-benzyl-D-glucono-1,5-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H34O6
CAS Number:
Molecular Weight:
538.63
Beilstein:
1276658
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

SMILES string

O=C1O[C@H](COCc2ccccc2)[C@@H](OCc3ccccc3)[C@H](OCc4ccccc4)[C@H]1OCc5ccccc5

InChI

1S/C34H34O6/c35-34-33(39-24-29-19-11-4-12-20-29)32(38-23-28-17-9-3-10-18-28)31(37-22-27-15-7-2-8-16-27)30(40-34)25-36-21-26-13-5-1-6-14-26/h1-20,30-33H,21-25H2/t30-,31-,32+,33-/m1/s1

InChI key

BUBVLQDEIIUIQG-NXVJRICRSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service