Skip to Content
Merck
All Photos(1)

Key Documents

687324

Sigma-Aldrich

N-Boc-cis-4-fluoro-L-proline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16FNO4
CAS Number:
Molecular Weight:
233.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]22/D −71.0±5°, c = 1 in chloroform

mp

157-161 °C (D)

functional group

carboxylic acid
fluoro

SMILES string

CC(C)(C)OC(=O)N1C[C@@H](F)C[C@H]1C(O)=O

InChI

1S/C10H16FNO4/c1-10(2,3)16-9(15)12-5-6(11)4-7(12)8(13)14/h6-7H,4-5H2,1-3H3,(H,13,14)/t6-,7-/m0/s1

InChI key

YGWZXQOYEBWUTH-BQBZGAKWSA-N

Application

N-Boc-cis-4-fluoro-L-proline can be used as a substrate in the preparation of:
  • α4β2 Receptor ligands bearing pyrrolidine nucleus.[1]
  • β-Amino pyrrolidine-2-carbonitrile derivatives as possible dipeptidyl peptidase IV (DPP4) inhibitors.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, synthesis and biological evaluation of 4-fluoropyrrolidine-2-carbonitrile and octahydrocyclopenta [b] pyrrole-2-carbonitrile derivatives as dipeptidyl peptidase IV inhibitors
Ji X, et al.
European Journal of Medicinal Chemistry, 86(6), 242-256 (2014)
Synthesis and Pharmacological Evaluation of α4β2 Nicotinic Ligands with a 3-Fluoropyrrolidine Nucleus
Tamborini L, et al.
ChemMedChem, 10(6), 1071-1078 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service