Skip to Content
Merck
All Photos(5)

Key Documents

685860

Sigma-Aldrich

(1R,2R)-1,2-Bis(2-hydroxyphenyl)ethylenediamine

95% (HPLC)

Synonym(s):

(1R,2R)-1,2-Bis(2-hydroxyphenyl)-1,2-ethanediamine, (R,R) Diaminopharm Diamine, (R,R) Jik Chin Mother Diamine, (R,R) Mother Diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O2
CAS Number:
Molecular Weight:
244.29
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 685860, please contact your local Merck office or dealer. Contact Technical Service

Assay

95% (HPLC)

form

powder

optical activity

[α]22/D +62°, c = 1% in chloroform

mp

157-163 °C

functional group

amine

SMILES string

N[C@@H]([C@H](N)c1ccccc1O)c2ccccc2O

InChI

1S/C14H16N2O2/c15-13(9-5-1-3-7-11(9)17)14(16)10-6-2-4-8-12(10)18/h1-8,13-14,17-18H,15-16H2/t13-,14-/m1/s1

InChI key

MRNPLGLZBUDMRE-ZIAGYGMSSA-N

General description

(1R, 2R)-1,2-Bis(2-hydroxyphenyl)ethylenediamine or (R, R)-1,2-bis(2-hydroxyphenyl)-1,2-diaminoethane (HPEN) is commonly used in the stereoselective synthesis of enantiopure compounds.[1]

Application

(1R, 2R)-1,2-Bis(2-hydroxyphenyl)ethylenediamine can be used as a precursor for the synthesis of:
  • Enantiomerically pure trans-3-arylpiperazine-2-carboxylic acid derivatives via diaza-Cope rearrangement (DCR).[1]
  • 4,4′-(1,2-Diazaniumylethane-1,2-diyl)dibenzoate trihydrate by treating with terephthalaldehydic acid.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4, 4?-(1, 2-Diazaniumylethane-1, 2-diyl) dibenzoate trihydrate
Numata T, et al.
IUCrData, 1(2), x160252-x160252 (2016)
Synthesis of Enantiopure Mixed Alkyl-Aryl Vicinal Diamines by the Diaza-Cope Rearrangement: A Synthesis of (+)-CP-99,994.
Kim, Miji and Kim, Hyeseung and Kim, Hyunwoo and Chin, Jik
The Journal of Organic Chemistry, 82(23), 12050-12058 (2017)
Short synthesis of enantiopure trans-3-arylpiperazine-2-carboxylic acid derivatives via diaza-Cope rearrangement
Kwon SH, et al.
Organic Letters, 14(14) (2012)
Stereospecific synthesis of C 2 symmetric diamines from the mother diamine by resonance-assisted hydrogen-bond directed diaza-Cope rearrangement.
Kim H, et al.
Journal of the American Chemical Society, 130(36), 12184-12191 (2008)
An efficient synthesis of chiral diamines with rigid backbones: Application in enantioselective michael addition of malonates to nitroalkenes.
Zhu, Qiming and Huang, Hanmin and Shi, Dengjian and Shen, Zhiqiang and Xia, Chungu
Organic Letters, 11(20), 4536-4539 (2009)

Articles

Chiral vicinal diamines are of tremendous interest to the synthetic chemist as they are found in many chiral catalysts and pharmaceuticals.

Related Content

The Chin group is interested in computational and experimental approaches to understanding stereoselective recognition and catalysis. Their studies in weak forces (H-bonding, electronic and steric effects) has led to a highly efficient method for making limitless varieties of chiral vicinal diamines from the 'mother diamine' that are useful for developing stereoselective organocatalysts or transition metal-based catalysts as well as for developing drugs (Acc Chem Res (2012) p1345). The 'mother diamine' is also useful for making binol, monophos and binap analogs. The Chin group is also interested in using reversible covalent bonds for stereoselective recognition and L to D conversion of natural and non-natural amino acids (EJOC (2012) p229).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service