Skip to Content
Merck
All Photos(1)

Key Documents

684317

Sigma-Aldrich

1-(Methyldithiocarbonyl)imidazole

96%

Synonym(s):

1H-Imidazole-1-carbodithioic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2S2
CAS Number:
Molecular Weight:
158.24
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

96%

refractive index

n20/D 1.6741

mp

29-33 °C

density

1.294 at 25 °C

SMILES string

CSC(=S)n1ccnc1

InChI

1S/C5H6N2S2/c1-9-5(8)7-3-2-6-4-7/h2-4H,1H3

InChI key

ZSKBYJIWBCCFMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Conversion of alcohols to O-alkyl-S-Methyldithiocarbonates, can be used to make thiourea and other dithiocarbonates.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 5, 675-678 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service