Skip to Content
Merck
All Photos(1)

Documents

678627

Sigma-Aldrich

3,4-Dichlorobenzylsulfonyl chloride

97%

Synonym(s):

(3,4-Dichlorophenyl)methanesulfonyl chloride, 3,4-Dichloro-α-toluenesulfonyl chloride, 3,4-Dichlorobenzenemethanesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5Cl3O2S
CAS Number:
Molecular Weight:
259.54
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

65-69 °C

SMILES string

Clc1ccc(CS(Cl)(=O)=O)cc1Cl

InChI

1S/C7H5Cl3O2S/c8-6-2-1-5(3-7(6)9)4-13(10,11)12/h1-3H,4H2

InChI key

DRYGYFAFCNDLEJ-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sulfonyl chlorides are often chosen as building blocks in medicinal chemistry for their ability to easily react with heterocyclic amines to create complex sulfonamides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service