Skip to Content
Merck
All Photos(1)

Key Documents

675024

Sigma-Aldrich

5-(4-Aminophenyl)-1,3,4-oxadiazole-2-thiol

97%

Synonym(s):

2-(4-Aminophenyl)-1,3,4-oxadiazole-5-thione, 5-(4-Aminophenyl)-1,3,4-oxadiazole-2(3H)-thione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3OS
CAS Number:
Molecular Weight:
193.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

235 °C (dec.)

SMILES string

Nc1ccc(cc1)-c2nnc(S)o2

InChI

1S/C8H7N3OS/c9-6-3-1-5(2-4-6)7-10-11-8(13)12-7/h1-4H,9H2,(H,11,13)

InChI key

FIGMOSDEYMCQLX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Davydova et al.
Journal of visualized experiments : JoVE, (145) (2019-03-26)
Maleimide-bearing bifunctional probes have been employed for decades for the site-selective modification of thiols in biomolecules, especially antibodies. Yet maleimide-based conjugates display limited stability in vivo because the succinimidyl thioether linkage can undergo a retro-Michael reaction. This, of course, can

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service