Skip to Content
Merck
All Photos(1)

Key Documents

674184

Sigma-Aldrich

S-Isopropyl chlorothioformate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHSCOCl
CAS Number:
Molecular Weight:
138.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

refractive index

n20/D 1.4738

bp

44-44 °C/20 mmHg

density

1.116 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)SC(Cl)=O

InChI

1S/C4H7ClOS/c1-3(2)7-4(5)6/h3H,1-2H3

InChI key

RQBXSTFDPJDJDH-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service