Skip to Content
Merck
All Photos(1)

Key Documents

673692

Sigma-Aldrich

4-Methyloxazole-5-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NO3
CAS Number:
Molecular Weight:
127.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

239-243 °C

functional group

carboxylic acid

SMILES string

Cc1ncoc1C(O)=O

InChI

1S/C5H5NO3/c1-3-4(5(7)8)9-2-6-3/h2H,1H3,(H,7,8)

InChI key

ZIXUNDOOBLSXPE-UHFFFAOYSA-N

Application

Used in a palladium-catalyzed cross-coupling between heteroaryl carboxylic acids and aryl bromides leading to arylated heterocycles.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pat Forgione et al.
Journal of the American Chemical Society, 128(35), 11350-11351 (2006-08-31)
Aryl-substituted five-membered heteroaromatics have attracted great interest over the past years due to their presence in a large number of pharmaceuticals and natural products. Recently, an advance in the preparation of these scaffolds was achieved by employing a C-H functionalization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service