Skip to Content
Merck
All Photos(1)

Key Documents

669075

Sigma-Aldrich

Dibutyltin diisothiocyanate

97%

Synonym(s):

NSC 292417, Dibutyldiisothiocyanato-stannane, Dibutyldiisothiocyanato-tin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18N2S2Sn
CAS Number:
Molecular Weight:
349.10
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

form

solid

reaction suitability

core: tin
reagent type: catalyst

mp

141-143 °C

SMILES string

CCCC[Sn](CCCC)(N=C=S)N=C=S

InChI

1S/2C4H9.2CNS.Sn/c2*1-3-4-2;2*2-1-3;/h2*1,3-4H2,2H3;;;/q;;2*-1;+2

InChI key

JVLGWEJONYVDMO-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Macrolactones
  • Isocyanate and isothiocyanate d1- and d2-complexes of vanadocene

Reactant for:
  • Unusual adduct formation with organotin(IV) Lewis acids
  • Interactions with vanadyl salicylaldimine β-ketimine and acetylacetonato complexes

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - STOT RE 1

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service