667935
Tungsten(VI) Ethoxide
99%
Synonym(s):
Hexaethoxytungsten
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
99%
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service