Skip to Content
Merck
All Photos(1)

Key Documents

667242

Sigma-Aldrich

Chloro(indenyl)bis(triphenylphosphine)ruthenium(II)

may contain ≤1 molar equivalent dichloromethane/acetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C9H7 · RuCl · 2(C18H15P)
CAS Number:
Molecular Weight:
776.25
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

impurities

may contain ≤1 molar equivalent dichloromethane/acetone

SMILES string

Cl[Ru].[CH]1[CH][C]2C=CC=C[C]2[CH]1.c3ccc(cc3)P(c4ccccc4)c5ccccc5.c6ccc(cc6)P(c7ccccc7)c8ccccc8

InChI

1S/2C18H15P.C9H7.ClH.Ru/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2-5-9-7-3-6-8(9)4-1;;/h2*1-15H;1-7H;1H;/q;;;;+1/p-1

InChI key

AUSVTIHCDLKYHV-UHFFFAOYSA-M

Application

Catalyst for olefination of carbonyl compounds.[1] Catalyst precursor for living radical polymerization.[2] Catalyst for redox isomerization of propargylic alcohols.[3]
Catalyst for:
  • Single-chain folding of polymers
  • Domino redox bicycloisomerization
  • Selective radical addition with a designed heterobifunctional halide
  • Tandem living radical polymerization via in situ monomer transformation with alcohols
  • Atom-economical synthesis of nitrogen heterocycles

Reactant for the synthesis of chiral-at-metal ruthenium allenylidene complexes

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pedro et al.
Organometallics, 26, 302-302 (2007)
Lizuka et al.
European Journal of Organic Chemistry, 782-782 (2007)
Trost, B.M. et al.
Journal of the American Chemical Society, ASAP-ASAP (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service