Skip to Content
Merck
All Photos(1)

Key Documents

652245

Sigma-Aldrich

2-((2′-Chloro-5′-(trifluoromethyl)phenoxy)methyl)phenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11BClF3O3
CAS Number:
Molecular Weight:
330.49
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥95%

form

solid

mp

138-147 °C (lit.)

SMILES string

OB(O)c1ccccc1COc2cc(ccc2Cl)C(F)(F)F

InChI

1S/C14H11BClF3O3/c16-12-6-5-10(14(17,18)19)7-13(12)22-8-9-3-1-2-4-11(9)15(20)21/h1-7,20-21H,8H2

InChI key

DNZSKCIEUYUZRA-UHFFFAOYSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service