About This Item
Recommended Products
Assay
97%
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings
refractive index
n20/D 1.538 (lit.)
density
0.986 g/mL at 20 °C (lit.)
0.979 g/mL at 25 °C
functional group
phosphine
SMILES string
C1CCC(C1)P(C2CCCC2)C3CCCC3
InChI
1S/C15H27P/c1-2-8-13(7-1)16(14-9-3-4-10-14)15-11-5-6-12-15/h13-15H,1-12H2
InChI key
DHWBYAACHDUFAT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Nickel-catalyzed cycloaddition of enones and alkynes
- Rhodium-catalyzed allylation of aldehydes with homoallylic alcohols
- Hydrocarbamoylation of alkynes by nickel/Lewis-acid catalysis
- Alkenylation of imidazoles by nickel/Lewis acid catalysis
- Palladium-catalyzed Negishi cross-coupling reactions
- PHosphine-catalyzed synthesis of 6-substituted 2-pyrones
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 3
Flash Point(F)
410.0 °F - closed cup
Flash Point(C)
210 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service