Skip to Content
Merck
All Photos(1)

Key Documents

647284

Sigma-Aldrich

2-Pyridineboronic acid N-phenyldiethanolamine ester

Synonym(s):

[[2,2′-(Phenylimino-κN)bis[ethanolato-κO]](2-)]-2-pyridinylboron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17BN2O2
CAS Number:
Molecular Weight:
268.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

>300 °C (lit.)

SMILES string

B1(C2=CC=CC=N2)OCCN(C3=CC=CC=C3)CCO1

InChI

1S/C15H17BN2O2/c1-2-6-14(7-3-1)18-10-12-19-16(20-13-11-18)15-8-4-5-9-17-15/h1-9H,10-13H2

InChI key

QDIQDVHNUYDVDD-UHFFFAOYSA-N

General description

May contain varying amounts of isopropanol and N-phenyldiethanolamine as stabilizers

Application

Reactant involved in Suzuki-Miyaura cross-coupling reactions for synthesis of 2-pyridylboronate

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hodgson, P.B.; Salingue, F.H.
Tetrahedron Letters, 685-687 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service