Skip to Content
Merck
All Photos(1)

Key Documents

643564

Sigma-Aldrich

3-(Trifluoromethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4F3N
CAS Number:
Molecular Weight:
147.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.418

bp

113-115 °C (lit.)

density

1.276 g/mL at 25 °C

functional group

fluoro

SMILES string

FC(F)(F)c1cccnc1

InChI

1S/C6H4F3N/c7-6(8,9)5-2-1-3-10-4-5/h1-4H

InChI key

JTZSFNHHVULOGJ-UHFFFAOYSA-N

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

73.4 °F - closed cup

Flash Point(C)

23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P M Hext et al.
Toxicology, 72(1), 61-75 (1992-01-01)
The accumulation of [methyl-14C]3-trifluoromethylpyridine (14C-3-FMP) by rat olfactory and hepatic tissue in vivo and in vitro has been investigated. 14C-3-FMP accumulates rapidly and selectively in both tissues in vivo, with an appreciable proportion of this activity being associated with the
B A Gaskell et al.
Toxicology, 62(1), 35-51 (1990-05-14)
The effects of a single exposure to 3-trifluoromethyl pyridine (3FMP), were investigated in two studies. In the first study, rats were exposed nose only to 0, 50 or 800 ppm 3FMP for periods of 15 min to 4 h. Half
B A Gaskell et al.
Toxicology, 50(1), 57-68 (1988-06-01)
Rats exposed by inhalation to 3-trifluoromethylpyridine (3-FMP) for 10, 30 or 90 days showed an unusual response in the nasal passages. Focal histological change including reduction in the number of cell layers, disorganisation, vacuolation and minimal necrosis was confined to
Malcolm E Tessensohn et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 18(16), 2250-2257 (2017-06-14)
The voltammetric behavior of 2,3,5,6-tetramethyl-1,4-phenylenediamine was found to be able to differentiate the hydrogen acceptor abilities of electroinactive pyridine compounds in acetonitrile. Weak and strong hydrogen acceptors were distinguished through the onset of a third oxidation process that came about

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service