Skip to Content
Merck
All Photos(1)

Key Documents

59855

Sigma-Aldrich

Isovaleric anhydride

≥95.0% (GC)

Synonym(s):

3-Methylbutyric anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2CO]2O
CAS Number:
Molecular Weight:
186.25
Beilstein:
1635850
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

refractive index

n20/D 1.417

density

0.932 g/mL at 20 °C (lit.)

SMILES string

CC(C)CC(=O)OC(=O)CC(C)C

InChI

1S/C10H18O3/c1-7(2)5-9(11)13-10(12)6-8(3)4/h7-8H,5-6H2,1-4H3

InChI key

FREZLSIGWNCSOQ-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service