Skip to Content
Merck
All Photos(1)

Key Documents

589144

Sigma-Aldrich

2-Ethylacryloyl chloride

contains 150 ppm BHT as inhibitor, 90%

Synonym(s):

α-Ethylacrylic acid chloride, 2-Methylenebutanoyl chloride, Ethacryloyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(=CH2)COCl
CAS Number:
Molecular Weight:
118.56
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

90%

contains

150 ppm BHT as inhibitor

refractive index

n20/D 1.444 (lit.)

bp

118-126 °C (lit.)

density

1.116 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

[H]\C([H])=C(\CC)C(Cl)=O

InChI

1S/C5H7ClO/c1-3-4(2)5(6)7/h2-3H2,1H3

InChI key

IIYDTSAAECYHAE-UHFFFAOYSA-N

General description

Acid chloride is a building block for organic synthesis.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

62.6 °F

Flash Point(C)

17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service