Skip to Content
Merck
All Photos(1)

Documents

579335

Sigma-Aldrich

5-Methoxy-2-tetralone

97%

Synonym(s):

5-methoxy-3,4-dihydronaphthalen-2(1H)-one, 5-Methoxy-3,4-dihydronaphthalen-2(1H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O2
CAS Number:
Molecular Weight:
176.21
Beilstein:
1451623
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

32-36 °C (lit.)

SMILES string

COc1cccc2CC(=O)CCc12

InChI

1S/C11H12O2/c1-13-11-4-2-3-8-7-9(12)5-6-10(8)11/h2-4H,5-7H2,1H3

InChI key

MDAIAXRTLTVEOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service