Skip to Content
Merck
All Photos(1)

Documents

578010

Sigma-Aldrich

4-Amino-3-iodobenzonitrile

98%

Synonym(s):

1-Amino-2-iodo-4-cyanobenzene, 4-Cyano-2-iodoaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H3(NH2)CN
CAS Number:
Molecular Weight:
244.03
MDL number:
UNSPSC Code:
12352100

Assay

98%

mp

112-115 °C (lit.)

SMILES string

Nc1ccc(cc1I)C#N

InChI

1S/C7H5IN2/c8-6-3-5(4-9)1-2-7(6)10/h1-3H,10H2

InChI key

UOWVTQFTEAYDLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Hydroamination-double hydroarylation for synthesis of fuzed carbazoles
  • Cycloisomerizations of aromatic homo- and bis-homopropargylic amine / amide intermediates
  • Asymmetric organocatalytic condensation and cycloaddition
  • Regioselective aryl C-H bond functionalization
  • Synthesis of [(triisopropylsilyl)ethynyl]indole derivatives
  • Domino condensation / S-arylation / heterocyclization reactions

Packaging

Packaged in glass bottles

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service