Skip to Content
Merck
All Photos(1)

Documents

572160

Sigma-Aldrich

4,4′-Ethylenedianiline

Synonym(s):

α,α′-Bi-p-toluidine, 4,4′-Diaminobibenzyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2
CAS Number:
Molecular Weight:
212.29
Beilstein:
394999
EC Number:
MDL number:
UNSPSC Code:
12162002

grade

technical grade

mp

133-137 °C (lit.)

SMILES string

Nc1ccc(CCc2ccc(N)cc2)cc1

InChI

1S/C14H16N2/c15-13-7-3-11(4-8-13)1-2-12-5-9-14(16)10-6-12/h3-10H,1-2,15-16H2

InChI key

UHNUHZHQLCGZDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service