Skip to Content
Merck
All Photos(1)

Key Documents

570907

Sigma-Aldrich

Benzyl (S,S,S)-2-azabicyclo[3.3.0]octane-3-carboxylate hydrochloride

97%

Synonym(s):

(S,S,S)-2-Azabicyclo-[3,3,0]-octane carboxylic acid benzylester hydrochloride, (S,S,S)-2-Azabicyclo[3,3,0]octane-3-carboxylic acid benzyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19NO2 ·HCl
CAS Number:
Molecular Weight:
281.78
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D -26°, c = 1% in DMSO

mp

176-180 °C (lit.)

functional group

ester
phenyl

SMILES string

Cl[H].[H][C@@]12CCC[C@]1([H])N[C@@H](C2)C(=O)OCc3ccccc3

InChI

1S/C15H19NO2.ClH/c17-15(18-10-11-5-2-1-3-6-11)14-9-12-7-4-8-13(12)16-14;/h1-3,5-6,12-14,16H,4,7-10H2;1H/t12-,13-,14-;/m0./s1

InChI key

HLXCXOQXUDRJLF-JKBZPBJLSA-N

Application

Benzyl (S,S,S)-2-azabicyclo[3.3.0]octane-3-carboxylate hydrochloride can be used:
  • In the preparation of ramipril, an angiotensin-converting enzyme (ACE) inhibitor.[1]
  • As a starting material for the synthesis of octahydrocyclopenta[b]pyrrole-2-carbonitrile derivatives as potent DPP4 inhibitors.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xun Ji et al.
European journal of medicinal chemistry, 86, 242-256 (2014-08-29)
Based on the previous work in our group and the principle of computer-aided drug design, a series of novel β-amino pyrrole-2-carbonitrile derivatives was designed and synthesized. Compounds 8l and 9l were efficacious and selective DPP4 inhibitors resulting in decreased blood
Expeditious synthesis of Ramipril: an Angiotensin-converting enzyme (ACE) inhibitor
Malakondaiah GC, et al.
Synthetic Communications, 38(11), 1737-1744 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service