570508
JandaJel™-Rink amide
100-200 mesh, extent of labeling: 0.5 mmol/g loading, 1 % cross-linked
Synonym(s):
4-[(2,4-Dimethoxyphenyl)Fmoc-amino)methyl]phenoxyacetamido methyl on JandaJel™, Knorr linker on JandaJel™
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
crosslinking
1 % cross-linked
extent of labeling
0.5 mmol/g loading
particle size
100-200 mesh
Legal Information
JandaJel is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service