Skip to Content
Merck
All Photos(1)

Key Documents

555398

Sigma-Aldrich

(4-Benzyloxy)phenylacetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4CH2CO2H
CAS Number:
Molecular Weight:
242.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

119-123 °C (lit.)

functional group

carboxylic acid
phenyl

SMILES string

OC(=O)Cc1ccc(OCc2ccccc2)cc1

InChI

1S/C15H14O3/c16-15(17)10-12-6-8-14(9-7-12)18-11-13-4-2-1-3-5-13/h1-9H,10-11H2,(H,16,17)

InChI key

XJHGAJLIKDAOPE-UHFFFAOYSA-N

General description

(4-Benzyloxy)phenylacetic acid is a para-substituted phenyl-acetic acid derivative.[1] Its crystals belong to the triclinic crystal system. Molecules of (4-benzyloxy)phenylacetic acid exists in centrosymmetric dimeric forms linked by double hydrogen bonds between carboxyl groups.[2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystallographic studies and physicochemical properties of Π-electron compounds. XVII. The structure of p-nitrophenylacetic acid.
Grabowski SJ, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 46(3), 428-430 (1990)
[4-(Benzyloxy) phenyl] acetic acid, C15H14O3.
Bats JW and Canenbley R.
Acta Crystallographica Section C, Crystal Structure Communications, 40(6), 993-995 (1984)
Tsung-I Hsu et al.
Oncotarget, 6(15), 13671-13687 (2015-04-25)
Herein, we evaluated the anti-cancer effect and molecular mechanisms of a novel betulinic acid (BA) derivative, SYK023, by using two mouse models of lung cancer driven by KrasG12D or EGFRL858R. We found that SYK023 inhibits lung tumor proliferation, without side

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service