All Photos(1)
About This Item
Linear Formula:
F3C6H2CH2Br
CAS Number:
Molecular Weight:
225.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
97%
refractive index
n20/D 1.5070 (lit.)
density
1.710 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
Fc1ccc(CBr)c(F)c1F
InChI
1S/C7H4BrF3/c8-3-4-1-2-5(9)7(11)6(4)10/h1-2H,3H2
InChI key
DGSXDQVPGXFOAN-UHFFFAOYSA-N
General description
2,3,4-Trifluorobenzyl bromide [1-(Bromomethyl)-2,3,4-trifluorobenzene] may be used to synthesize phase-transfer catalysts (PTCs).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
195.1 °F - closed cup
Flash Point(C)
90.6 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of (αR, βS)-epoxyketones by asymmetric epoxidation of chalcones with cinchona phase-transfer catalysts.
Yoo MS, et al.
Tetrahedron Letters, 51(42), 5601-5603 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service