Skip to Content
Merck
All Photos(2)

Key Documents

552402

Sigma-Aldrich

1-Bis(4-fluorophenyl)methyl piperazine

97%

Synonym(s):

1-(4,4′-Difluorobenzhydryl)piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18F2N2
CAS Number:
Molecular Weight:
288.34
Beilstein:
620754
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

88-92 °C (lit.)

SMILES string

Fc1ccc(cc1)C(N2CCNCC2)c3ccc(F)cc3

InChI

1S/C17H18F2N2/c18-15-5-1-13(2-6-15)17(21-11-9-20-10-12-21)14-3-7-16(19)8-4-14/h1-8,17,20H,9-12H2

InChI key

TTXIFFYPVGWLSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bis(4-fluorophenyl)methyl piperazine is formed during the synthesis of flunarizine. Its crystals exhibits monoclinic crystal system and space group P21/c.[1]

Application

1-Bis(4-fluorophenyl)methyl piperazine may be used to synthesize 1-[bis(4-fluorophenyl)methyl]-4-[3-[(N-ethoxycarbonyl-N-phenyl)amino]-2-hydroxypropyl]piperazine[2] and (R, S) 3-[4-(bis(4-fluorophenyl)methyl)piperazin-1-yl]dihydrofuran-2(3H)-one.[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and biological evaluation of new derivatives of 2-substituted 4-hydroxybutanamides as GABA uptake inhibitors
Kulig K, et al.
European Journal of Medicinal Chemistry, 46(1), 183-190 (2011)
Syntheses of novel diphenyl piperazine derivatives and their activities as inhibitors of dopamine uptake in the central nervous system.
Kimura M, et al.
Bioorganic & Medicinal Chemistry, 11(8), 1621-1630 (2003)
1-[Bis (4-fluorophenyl) methyl] piperazine.
Dayananda AS, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 68(9), o2817-o2817 (2012)
Y Yamanaka et al.
Archives of toxicology, 69(6), 391-396 (1995-01-01)
Toxic effects of a detriazinyl metabolite of almitrine (DTMA) were evaluated in rats and on cultured rat macrophages. In rats daily treated with DTMA for 16 weeks, spastic gaits with heel-lifting appeared, and lamellated and/or crystalloid bodies formed in sensory
S Kariya et al.
Research communications in chemical pathology and pharmacology, 78(1), 85-95 (1992-10-01)
The oxidative metabolism of flunarizine [1-[bis(4-fluorophenyl)-methyl]-4-(3-phenyl-2-propenyl)piperazine, FZ] to 1-[bis-(4-fluorophenyl)methyl]piperazine (M-1), 1-[bis(4-fluorophenyl)methyl]-4-[3-(4'-hydroxyphenyl)-2- propenyl]piperazine (M-2) and 4,4'-difluorobenzophenone (M-3) has been studied in liver microsomes of Wistar and Dark Agouti (DA) rats. Kinetic analysis demonstrated a sex difference (male > female) in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service