Skip to Content
Merck
All Photos(2)

Key Documents

552054

Sigma-Aldrich

Cyclotriphosphazene-PMMH-6 dendrimer, generation 1.0

Synonym(s):

Cyclotriphosphazene-phenoxymethyl(methylhydrazono) dendrimer, generation 1.0

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N3P3(OC6H4CHNN(CH3)P(S)Cl2)6
CAS Number:
Molecular Weight:
1827.58
MDL number:
UNSPSC Code:
12162002
Pricing and availability is not currently available.

description

dichlorophosphinothioyl surface groups

form

powder

mol wt

generation 1

feature

core type Phosphorous dendrimers with cyclotriphosphazene core

no. Surface Groups

6

pH

1.01 (5 wt. % in ethanol)

mp

117-120.8 °C (lit.)

General description

Contains 6 surface dichlorophosphinothioyl groups.

Application

Stable up to 200 °C, high solvent solubility, air stable, high values of dipole moments (10 - 300 Debyes)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service