Skip to Content
Merck
All Photos(1)

Key Documents

549800

Sigma-Aldrich

4-Bromo-2,3-dimethylanisole

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH3)2C6H2OCH3
CAS Number:
Molecular Weight:
215.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

refractive index

n20/D 1.566 (lit.)

bp

197 °C (lit.)

density

1.36 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(Br)c(C)c1C

InChI

1S/C9H11BrO/c1-6-7(2)9(11-3)5-4-8(6)10/h4-5H,1-3H3

InChI key

WOWNZZYJCUUIFC-UHFFFAOYSA-N

General description

4-Bromo-2,3-dimethylanisole can be prepared from 2,3-dimethylanisole via bromination using N-bromosuccinimide-acetonitrile (NBS-CH3CN) system.

Application

4-Bromo-2,3-dimethylanisole may be used in the preparation of:
  • 4,6-Dibromo-2,3-dimethylanisole via bromination using NBS-CH3CN system.
  • 4-Methoxy-2,2′,3,3′-tetramethyldiphenyl ether by reacting with 2,3-dimethylphenol.
  • A novel arylsulfonamide derivative with potent glucocorticoid receptor (GR) agonist activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-Bromosuccinimide in acetonitrile: A mild and regiospecific nuclear brominating reagent for methoxybenzenes and naphthalenes.
Carreno MC, et al.
The Journal of Organic Chemistry, 60(16), 5328-5331 (1995)
Oxidative coupling of phenols. Part 9. The role of steric effects in the oxidation of methyl-substituted phenols.
Armstrong DR, et al.
J. Chem. Soc. Perkin Trans. II, 5, 581-585 (1983)
Substituted phenyl as a steroid A-ring mimetic: Providing agonist activity to a class of arylsulfonamide nonsteroidal glucocorticoid ligands.
Darren D, et al.
Bioorganic & Medicinal Chemistry Letters, 23(24), 6645-6649 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service