Skip to Content
Merck
All Photos(1)

Key Documents

546755

Sigma-Aldrich

2-Bromo-1,4-bis(2-ethylhexyloxy)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3CH(CH2CH3)CH2O]C6H3Br
CAS Number:
Molecular Weight:
413.43
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

refractive index

n20/D 1.506 (lit.)

bp

200-225 °C/2 mmHg (lit.)

density

1.114 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)COc1ccc(OCC(CC)CCCC)c(Br)c1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service