Skip to Content
Merck
All Photos(2)

Key Documents

545899

Sigma-Aldrich

3,5-Dichlorocatechol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(OH)2
CAS Number:
Molecular Weight:
179.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

79.5-84.5 °C (lit.)

SMILES string

Oc1cc(Cl)cc(Cl)c1O

InChI

1S/C6H4Cl2O2/c7-3-1-4(8)6(10)5(9)2-3/h1-2,9-10H

InChI key

XSXYVLIPQMXCBV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Liu et al.
Gene, 268(1-2), 207-214 (2001-05-23)
The modified-ortho pathway genes responsible for the degradation of chlorocatechols produced from 3- and 4-chlorobenzoate in Burkholderia sp. NK8 were cloned and analyzed. The five genes predicted to encode a LysR-type transcriptional regulator, chlorocatechol 1,2-dioxygenase, chloromuconate cycloisomerase, dienelactone hydrolase, and
Dirk Benndorf et al.
Proteomics, 6(11), 3319-3329 (2006-04-26)
Pseudomonas putida KT2440 is often used as a model to investigate toxicity mechanisms and adaptation to hazardous chemicals in bacteria. The objective of this paper was to test the impact of the chlorophenoxy herbicides 2,4-dichlorophenoxyacetic acid (2,4-D) and 2-(2,4-dichlorophenoxy)propanoic acid
E J Perkins et al.
Journal of bacteriology, 172(5), 2351-2359 (1990-05-01)
Growth of Alcaligenes eutrophus JMP134 on 2,4-dichlorophenoxyacetate requires a 2,4-dichlorphenol hydroxylase encoded by gene tfdB. Catabolism of either 2,4-dichlorophenoxyacetate or 3-chlorobenzoate involves enzymes encoded by the chlorocatechol oxidative operon consisting of tfdCDEF, which converts 3-chloro- and 3,5-dichlorocatechol to maleylacetate and
S N Gorlatov et al.
Journal of basic microbiology, 32(3), 177-184 (1992-01-01)
Effects of aromatic and nonaromatic cosubstrates on the rate of 2,4-dichlorophenol degradation by R. erythropolis 1CP were studied under growth and nongrowth conditions. Glucose and maltose were found to accelerate 2,4-dichlorophenol (initial conc. 50 mg/l) dechlorination from 11 days to
Stefan G Huber et al.
Environmental science & technology, 41(22), 7802-7806 (2007-12-14)
The formation of volatile compounds during abiotic degradation processes of aromatic compounds in soil has been the subject of many experimental studies but should be examined further. In this context, the present work investigates the natural formation of carbon suboxide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service