Skip to Content
Merck
All Photos(1)

Key Documents

543039

Sigma-Aldrich

(R)-1,4-Dioxaspiro[4.5]decane-2-carboxaldehyde

Synonym(s):

(R)-2,3-Cyclohexylideneglyceraldehyde, (R)-Cyclohexylideneglyceraldehyde, 2,3-O-Cyclohexylidene-D-glyceraldehyde, O,O-Cyclohexylidene-(R)-glyceraldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
₹13,985.90

₹13,985.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
₹13,985.90

About This Item

Empirical Formula (Hill Notation):
C9H14O3
CAS Number:
Molecular Weight:
170.21
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

₹13,985.90


Please contact Customer Service for Availability

Request a Bulk Order

optical activity

[α]/D 60±10° in chloroform

Quality Level

refractive index

n20/D 1.472 (lit.)

density

1.108 g/mL at 25 °C (lit.)

functional group

aldehyde
ether
ketal

SMILES string

O=C[C@H]1COC2(CCCCC2)O1

InChI

1S/C9H14O3/c10-6-8-7-11-9(12-8)4-2-1-3-5-9/h6,8H,1-5,7H2/t8-/m0/s1

InChI key

FFGZPNNLXMQFMO-QMMMGPOBSA-N

Application

Reactant for:
  • Proline-catalyzed Diels Alder reaction
  • Stereodivergent synthesis of carbahexofuranoses employing a Wittig olefination-Claisen rearrangement protocol
  • Wittig reactions
  • Crotylation reactions
  • Asymmetric synthesis of Goniothalesdiol A via stereocontrolled allylation and base-catalyzed oxy-Michael addition
Used as a building block.[1]

Other Notes

Contains varying amounts of polymer

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 60, 585-587 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service