Skip to Content
Merck
All Photos(1)

Key Documents

538450

Sigma-Aldrich

2-Chlorobenzenesulfonamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4SO2NH2
CAS Number:
Molecular Weight:
191.64
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

189-193 °C (lit.)

SMILES string

NS(=O)(=O)c1ccccc1Cl

InChI

1S/C6H6ClNO2S/c7-5-3-1-2-4-6(5)11(8,9)10/h1-4H,(H2,8,9,10)

InChI key

JCCBZCMSYUSCFM-UHFFFAOYSA-N

General description

2-Chlorobenzenesulfonamide is a halogenated sulfonamide derivative. It is formed as one of the major degradation product of chlorosulfuron. Crystals of 2-chlorobenzenesulfonamide exhibit monoclinic space group Cc. Its molecules are linked by N1—H11···O1 and N1—H12···O2 hydrogen bonding to form an infinite 3-D molecular network.

Application

2-Chlorobenzenesulfonamide may be used in the synthesis of 2-chloro-N-(4-methoxybenzoyl)benzenesulfonamide and 1,2,4-thiadiazine ,1-dioxides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fate of chlorsulfuron in the environment. 2. Field evaluations.
Strek HJ.
Pestic. Sci., 53(1), 52-70 (1998)
H P Hershey et al.
Plant molecular biology, 17(4), 679-690 (1991-10-01)
A search of compounds capable of inducing specific gene expression in plants without affecting growth and development led to the examination of changes in the pattern of gene expression in corn after treatment with substituted benzenesulfonamide herbicide safeners. Following hydroponic
Synthesis and biological activity of novel 3-heteroaryl-2H-pyrido [4, 3-e][1, 2, 4] thiadiazine and 3-heteroaryl-2H-benzo [e][1, 2, 4] thiadiazine 1, 1-dioxides.
Gobis, Katarzyna, et al.
Monatshefte fur Chemie / Chemical Monthly, 144(8), 1197-1203 (2013)
2-Chlorobenzenesulfonamide.
Gowda BT, et al.
Acta Crystallographica Section E, Structure Reports Online, 65(9), o2144-o2144 (2009)
Aldo Pagano et al.
PLoS genetics, 3(2), e1-e1 (2007-02-06)
By means of a computer search for upstream promoter elements (distal sequence element and proximal sequence element) typical of small nuclear RNA genes, we have identified in the human genome a number of previously unrecognized, putative transcription units whose predicted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service