Recommended Products
Assay
97%
refractive index
n20/D 1.518 (lit.)
bp
258 °C (lit.)
density
0.94 g/mL at 25 °C (lit.)
SMILES string
CC(C)(C)c1ccc(cc1)C#N
InChI
1S/C11H13N/c1-11(2,3)10-6-4-9(8-12)5-7-10/h4-7H,1-3H3
InChI key
IIZURLNRIMKEDL-UHFFFAOYSA-N
General description
4-tert-Butylbenzonitrile can be prepared from 4-tert-butyl-1,2-dihydrobenzene, via dehydrogenation.
Application
4-tert-Butylbenzonitrile may be used to synthesize 3-bromo-4′-tert-butylbenzophenone.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Allylic rearrangement of cyanophosphate. II Synthesis of ?-cyano-a,?-unsaturated ketones.
Chemical & Pharmaceutical Bulletin, 34(7), 2747-2753 (1986)
Synthesis of 1, 3-connected polyarylmethanes.
The Journal of Organic Chemistry, 56(7), 2557-2563 (1991)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service