Skip to Content
Merck
All Photos(1)

Key Documents

520047

Sigma-Aldrich

1-Ethynyl-4-fluorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4C≡CH
CAS Number:
Molecular Weight:
120.12
Beilstein:
1099285
MDL number:
UNSPSC Code:
12352100

Assay

98%

refractive index

n20/D 1.516 (lit.)

bp

55-56 °C/40 mmHg (lit.)

mp

26-27 °C (lit.)

density

1.048 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(cc1)C#C

InChI

1S/C8H5F/c1-2-7-3-5-8(9)6-4-7/h1,3-6H

InChI key

QXSWHQGIEKUBAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Manufactured by CB Research and Development

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service