Skip to Content
Merck
All Photos(1)

Key Documents

50807

Sigma-Aldrich

1,2-Dimethyl-3-propylimidazolium bis(trifluoromethylsulfonyl)imide

≥97.0% (HPLC)

Synonym(s):

DMPIIm

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15F6N3O4S2
CAS Number:
Molecular Weight:
419.36
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

form

liquid

impurities

<0.5% water

refractive index

n20/D 1.433

mp

15 °C (lit.)

density

1.436 g/mL at 25 °C

SMILES string

CCC[n+]1ccn(C)c1C.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C8H15N2.C2F6NO4S2/c1-4-5-10-7-6-9(3)8(10)2;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h6-7H,4-5H2,1-3H3;/q+1;-1

InChI key

XOZHIVUWCICHSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical form

Fluorinated, hydrophobic ionic liquid

Other Notes

Exremely hydrophobic ionic liquid. Thermal properties of imidazolium ionic liquids[1]; interfacial stability of Li in solvent-free ionic liquid[2]

Legal Information

Product protected by U.S. Patent 5,827,602 assigned to Covalent Associates, Inc. Woburn, Ma, USA

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H.L. Ngo et al.
Thermochimica Acta, 357, 97-97 (2000)
V.R. Koch et al.
Journal of the Electrochemical Society, 142, 116-116 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service