Skip to Content
Merck
All Photos(1)

Key Documents

496200

Sigma-Aldrich

3-Methyl-1-butanol

biotech. grade, ≥99%

Synonym(s):

Isoamyl alcohol, Isopentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1718835
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:

grade

biotech. grade

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

644 °F

expl. lim.

1.2-9 %, 100 °F

technique(s)

GC/MS: suitable

impurities

≤0.002 meq/g Titr. acid
≤0.005% (water)
≤0.1% carbonyl

evapn. residue

<0.0005%

color

APHA: <20

refractive index

n20/D 1.406 (lit.)

pH

5.6 (20 °C, 25 g/L)

bp

130 °C (lit.)

mp

−117 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCO

InChI

1S/C5H12O/c1-5(2)3-4-6/h5-6H,3-4H2,1-2H3

InChI key

PHTQWCKDNZKARW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methyl-1-butanol was used as standard in GC-MS analysis of volatile compounds produced by Rhizobacteria and Spanish Honeys.

Biochem/physiol Actions

3-Methyl-1-butanol is produced as a fusel alcohol in yeast fermentation. It is a naturally-produced fuel substitute that has advantages of acting as precursor for isoamyl acetate and replacement for gasoline.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

110.3 °F - closed cup

Flash Point(C)

43.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed A Farag et al.
Phytochemistry, 67(20), 2262-2268 (2006-09-05)
Chemical and plant growth studies of Bacilli strains GB03 and IN937a revealed that the volatile components 2,3-butanediol and acetoin trigger plant growth promotion in Arabidopsis. Differences in growth promotion when cytokinin-signaling mutants are exposed to GB03 versus IN937a volatiles suggest
Michael R Connor et al.
Applied and environmental microbiology, 74(18), 5769-5775 (2008-08-05)
3-Methyl-1-butanol is a potential fuel additive or substitute. Previously this compound was identified in small quantities in yeast fermentation as one of the fusel alcohols. In this work, we engineered an Escherichia coli strain to produce 3-methyl-1-butanol from glucose via
Niels O Verhulst et al.
Malaria journal, 10, 28-28 (2011-02-10)
Anopheles gambiae sensu stricto is considered to be highly anthropophilic and volatiles of human origin provide essential cues during its host-seeking behaviour. A synthetic blend of three human-derived volatiles, ammonia, lactic acid and tetradecanoic acid, attracts A. gambiae. In addition
Luca Bianco et al.
PloS one, 9(10), e110377-e110377 (2014-10-11)
High-density SNP arrays for genome-wide assessment of allelic variation have made high resolution genetic characterization of crop germplasm feasible. A medium density array for apple, the IRSC 8K SNP array, has been successfully developed and used for screens of bi-parental
Mary J Dunlop et al.
Molecular systems biology, 7, 487-487 (2011-05-11)
Many compounds being considered as candidates for advanced biofuels are toxic to microorganisms. This introduces an undesirable trade-off when engineering metabolic pathways for biofuel production because the engineered microbes must balance production against survival. Cellular export systems, such as efflux

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service