Skip to Content
Merck
All Photos(1)

Documents

493503

Sigma-Aldrich

Vinyl-d3 bromide

≥98 atom % D, ≥99% (CP), contains hydroquinone as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D2C=CDBr
CAS Number:
Molecular Weight:
109.97
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

vapor density

3.8 (vs air)

Quality Level

vapor pressure

1551 mmHg ( 37.8 °C)

isotopic purity

≥98 atom % D

Assay

≥99% (CP)

autoignition temp.

986 °F

contains

hydroquinone as stabilizer

refractive index

n20/D 1.435 (lit.)

bp

16 °C/750 mmHg (lit.)

mp

-139 °C (lit.)

density

1.56 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]\C([2H])=C(/[2H])Br

InChI

1S/C2H3Br/c1-2-3/h2H,1H2/i1D2,2D

InChI key

INLLPKCGLOXCIV-FUDHJZNOSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service