Skip to Content
Merck
All Photos(1)

Key Documents

481785

Sigma-Aldrich

4-Chloro-3-(trifluoromethyl)benzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CF3)CHO
CAS Number:
Molecular Weight:
208.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.504 (lit.)

bp

218 °C (lit.)

density

1.45 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1cc(C=O)ccc1Cl

InChI

1S/C8H4ClF3O/c9-7-2-1-5(4-13)3-6(7)8(10,11)12/h1-4H

InChI key

NIHMMULLFBKTOK-UHFFFAOYSA-N

General description

4-Chloro-3-(trifluoromethyl)benzaldehyde is a benzaldehyde derivative. Its enthalpy of vaporization at boiling point has been reported.

Application

4-Chloro-3-(trifluoromethyl)benzaldehyde may be used in the synthesis of 3′-[(3-trifluoromethyl-4-chloro)-benzylamino]-3′-deoxy-5′-O-triphenylmethyl-β-D-thymidine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, modeling and evaluation of 3'-(1-aryl-1H-tetrazol-5-ylamino)-substituted 3'-deoxythymidine derivatives as potent and selective human mitochondrial thymidine kinase inhibitors.
Van PS, et al.
Organic & Biomolecular Chemistry, 9(3), 892-901 (2011)
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 548-548 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service