Skip to Content
Merck
All Photos(1)

Key Documents

480495

Sigma-Aldrich

1,3-Bis(4-methoxyphenyl)-1,3-propanedione

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OC6H4CO)2CH2
CAS Number:
Molecular Weight:
284.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

108-115 °C (lit.)

SMILES string

COc1ccc(cc1)C(=O)CC(=O)c2ccc(OC)cc2

InChI

1S/C17H16O4/c1-20-14-7-3-12(4-8-14)16(18)11-17(19)13-5-9-15(21-2)10-6-13/h3-10H,11H2,1-2H3

InChI key

GNMDORSUZRRMFS-UHFFFAOYSA-N

General description

1,3-Bis(4-methoxyphenyl)-1,3-propanedione is a diketone. It is formed as an intermediate during the synthesis of bisphenols.[1]

Application

1,3-Bis(4-methoxyphenyl)-1,3-propanedione (Hpmdbm) may be used:
  • As a reactant in the preparation of 1,3-bis(4-hydroxyphenyl)-1,3-propanedione.[2]
  • As a starting material in the synthesis of propylpyrazole triol (PPT).[3]
  • As a ligand in the synthesis of [NaFe6(OCH3)12(pmdbm)6]ClO4, an iron(III)-oxo cluster.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James J Devery et al.
Synlett : accounts and rapid communications in synthetic organic chemistry, (9)(9), 1490-1494 (2009-01-01)
A convenient approach for the synthesis of tetrasubstituted pyrazoles is described. The method involves the treatment of 1,3-diketones and allyltrimethylsilane with CAN followed by cerium-catalyzed addition of substituted hydrazines to construct pyrazoles in good yields.
Thermostable and heat resistant aromatic polyethers based on heterocyclic bisphenols.
Rusanov AL and Belomoina NM.
Polymer Science, Series C, 51(1), 87-125 (2009)
Single-Ion versus Dipolar Origin of the Magnetic Anisotropy in Iron (III)-Oxo Clusters: A Case Study.
Abbati GL, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 7(8), 1796-1807 (2001)
β-Diketone reagents for the determination of borate in water.
Lambert JL, et al.
Analytical Chemistry, 50(6), 820-822 (1978)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service