Skip to Content
Merck
All Photos(1)

Documents

475661

Sigma-Aldrich

Trimethylolpropane benzoate diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCO2CH2)2C(C2H5)CH2O2CC6H5
CAS Number:
Molecular Weight:
346.37
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

contains

1,000 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.511 (lit.)

density

1.126 g/mL at 25 °C (lit.)

SMILES string

CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)c1ccccc1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service